siRNA Details
| Gene Symbol: Pdilt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013902.1 | NP_001013924.1 | 11, 12 | 1725 | ||
| XM_008759714.1 | XP_008757936.1 | 10, 11 | 1544 | |||
| GenBank mRNA | BC083897.1 | - | 11, 12 | 1725 | 7133354 | |

Pdilt
Genomic Map