siRNA Details
| Gene Symbol: Gpr139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024241.1 | NP_001019412.1 | 2 | 998 | ||
| GenBank mRNA | AB196531.1 | - | 2 | 998 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr139| Gene Symbol: Gpr139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024241.1 | NP_001019412.1 | 2 | 998 | ||
| GenBank mRNA | AB196531.1 | - | 2 | 998 | ||