siRNA Details
| Gene Symbol: Olr300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000237.1 | NP_001000237.1 | 1 | 427 | ||
| XM_008759976.1 | XP_008758198.1 | 3 | 2407 | |||
| XM_008759977.1 | XP_008758199.1 | 3 | 2534 | |||
| XM_008759978.1 | XP_008758200.1 | 3 | 2496 | |||

Olr300
Genomic Map