siRNA Details
| Gene Symbol: Ifitm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106314.1 | NP_001099784.1 | 3 | 433 | ||
| XM_006230530.1 | XP_006230592.1 | 3 | 516 | |||
| GenBank mRNA | FQ221702.1 | - | 2 | 286 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifitm1| Gene Symbol: Ifitm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106314.1 | NP_001099784.1 | 3 | 433 | ||
| XM_006230530.1 | XP_006230592.1 | 3 | 516 | |||
| GenBank mRNA | FQ221702.1 | - | 2 | 286 | ||