siRNA Details
| Gene Symbol: Brsk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760049.1 | XP_008758271.1 | 3 | 236 | ||
| GenBank mRNA | AM040976.1 | - | 488 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brsk2| Gene Symbol: Brsk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760049.1 | XP_008758271.1 | 3 | 236 | ||
| GenBank mRNA | AM040976.1 | - | 488 | |||