siRNA Details
| Gene Symbol: Tmem171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013903.1 | NP_001013925.1 | 4 | 1015 | ||
| XM_006231833.2 | XP_006231895.1 | 4 | 1932 | |||
| XM_006231834.2 | XP_006231896.1 | 4 | 1928 | |||
| XM_008760660.1 | XP_008758882.1 | 4 | 1448 | |||
| GenBank mRNA | AY185507.1 | - | 4 | 1015 | ||

Tmem171
Genomic Map