siRNA Details
| Gene Symbol: Tcirg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199089.2 | NP_954520.2 | 12 | 1501 | ||
| XM_006230727.2 | XP_006230789.1 | 13 | 1605 | |||
| XM_006230728.2 | XP_006230790.1 | 13 | 1576 | |||
| XM_008760097.1 | XP_008758319.1 | 13 | 1672 | |||
| GenBank mRNA | BC061859.1 | - | 13 | 1574 | 5598499 | |
| DQ286426.1 | - | 12 | 1501 | |||

Tcirg1
Genomic Map