siRNA Details
| Gene Symbol: Ndufv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006972.1 | NP_001006973.1 | 4, 5 | 520 | ||
| GenBank mRNA | BC083709.1 | - | 4, 5 | 520 | 7190583 | |
| FQ214688.1 | - | 4, 5 | 527 | |||
| FQ217076.1 | - | 4, 5 | 577 | |||

Ndufv1
Genomic Map