siRNA Details
| Gene Symbol: RGD1307603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134508.1 | NP_001127980.1 | 6 | 550 | ||
| XM_006230736.1 | XP_006230798.1 | 7 | 653 | |||
| XM_006230737.1 | XP_006230799.1 | 7 | 641 | |||
| XM_008760100.1 | XP_008758322.1 | 7 | 712 | |||
| GenBank mRNA | FQ219091.1 | - | 7 | 546 | ||

RGD1307603
Genomic Map