siRNA Details
| Gene Symbol: Zdhhc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039100.1 | NP_001034189.1 | 3 | 612 | ||
| GenBank mRNA | AY886540.1 | - | 3 | 612 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zdhhc24| Gene Symbol: Zdhhc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039100.1 | NP_001034189.1 | 3 | 612 | ||
| GenBank mRNA | AY886540.1 | - | 3 | 612 | ||