siRNA Details
| Gene Symbol: mrpl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006973.1 | NP_001006974.1 | 2 | 198 | ||
| XM_006230739.2 | XP_006230801.1 | 2 | 242 | |||
| GenBank mRNA | BC083740.1 | - | 2 | 198 | 7191782 | |
| FQ225172.1 | - | 2 | 223 | |||

mrpl11
Genomic Map