siRNA Details
Gene Symbol: LOC100911882 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749039.3 | XP_003749087.1 | 1 | 1699 |
Gene Symbol: Cd248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106325.1 | NP_001099795.1 | 1708 |
Gene Symbol: LOC100911932 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749040.3 | XP_003749088.1 | 1699 |
