siRNA Details
| Gene Symbol: VEGFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243733.1 | NP_001230662.1 | 7 | 1210 | ||
| NM_003377.4 | NP_003368.1 | 7 | 1311 | |||
| GenBank mRNA | BC008818.2 | - | 7 | 1101 | 4053976 | |
| BG031449.1 | - | 2 | 535 | |||
| BM694361.1 | - | 1 | 152 | |||
| U43368.1 | - | 7 | 1063 | |||

VEGFB
Genomic Map