siRNA Details
| Gene Symbol: Ctsw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024242.1 | NP_001019413.1 | 2 | 151 | ||
| GenBank mRNA | BC093401.1 | - | 2 | 151 | 7389584 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsw| Gene Symbol: Ctsw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024242.1 | NP_001019413.1 | 2 | 151 | ||
| GenBank mRNA | BC093401.1 | - | 2 | 151 | 7389584 | |