siRNA Details
| Gene Symbol: Efemp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005907.2 | NP_001005907.1 | 4 | 414 | ||
| NM_001277341.1 | NP_001264270.1 | 4 | 414 | |||
| XM_006230742.2 | XP_006230804.1 | 4 | 754 | |||
| XM_006230743.1 | XP_006230805.1 | 4 | 405 | |||
| XM_006230744.2 | XP_006230806.1 | 4 | 411 | |||
| GenBank mRNA | BC083804.1 | - | 4 | 352 | 7109182 | |
| FM037652.1 | - | 4 | 376 | |||

Efemp2
Genomic Map