siRNA Details
| Gene Symbol: Snx15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024752.1 | NP_001019923.1 | 1 | 165 | ||
| GenBank mRNA | BC097481.1 | - | 1 | 165 | 7459091 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx15| Gene Symbol: Snx15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024752.1 | NP_001019923.1 | 1 | 165 | ||
| GenBank mRNA | BC097481.1 | - | 1 | 165 | 7459091 | |