siRNA Details
| Gene Symbol: Nudt22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199090.2 | NP_954521.1 | 2 | 304 | ||
| XM_006230760.2 | XP_006230822.1 | 1 | 206 | |||
| GenBank mRNA | BC060593.1 | - | 2 | 305 | 6887766 | |
| FQ212766.1 | - | 2 | 251 | |||
| FQ215165.1 | - | 2 | 232 | |||
| FQ219356.1 | - | 2 | 255 | |||

Nudt22
Genomic Map