siRNA Details
| Gene Symbol: Incenp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106335.1 | NP_001099805.1 | 12 | 1905 | ||
| XM_006231024.1 | XP_006231086.1 | 13 | 1876 | |||
| XM_006231025.1 | XP_006231087.1 | 12 | 1864 | |||
| XM_006231026.1 | XP_006231088.1 | 13 | 1876 | |||
| XM_006231027.2 | XP_006231089.1 | 13 | 2640 | |||
| XM_006231028.2 | XP_006231090.1 | 13 | 1875 | |||

Incenp
Genomic Map