siRNA Details
| Gene Symbol: Best1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011940.1 | NP_001011940.1 | 10 | 1282 | ||
| XM_006231029.2 | XP_006231091.1 | 10 | 1322 | |||
| GenBank mRNA | BC079048.1 | - | 10 | 1282 | 7115295 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Best1| Gene Symbol: Best1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011940.1 | NP_001011940.1 | 10 | 1282 | ||
| XM_006231029.2 | XP_006231091.1 | 10 | 1322 | |||
| GenBank mRNA | BC079048.1 | - | 10 | 1282 | 7115295 | |