siRNA Details
| Gene Symbol: Myrf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170487.1 | NP_001163958.1 | 23, 24 | 3100 | ||
| XM_006231030.2 | XP_006231092.1 | 23, 24 | 3258 | |||
| XM_006231031.2 | XP_006231093.1 | 22, 23 | 3180 | |||
| XM_008760220.1 | XP_008758442.1 | 22, 23 | 3177 | |||
| XM_008760221.1 | XP_008758443.1 | 22, 23 | 2997 | |||
| XM_008760222.1 | XP_008758444.1 | 20, 21 | 2821 | |||

Myrf
Genomic Map