siRNA Details
| Gene Symbol: Ccdc86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006974.2 | NP_001006975.1 | 1 | 357 | ||
| GenBank mRNA | BC083770.1 | - | 1 | 375 | 7193350 | |
| DQ501253.1 | - | 1 | 357 | |||
| FQ213715.1 | - | 1 | 396 | |||
| FQ219684.1 | - | 1 | 406 | |||

Ccdc86
Genomic Map