siRNA Details
| Gene Symbol: Ms4a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181082.2 | NP_851596.1 | 6 | 892 | ||
| GenBank mRNA | AY258290.1 | - | 6 | 892 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ms4a14| Gene Symbol: Ms4a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181082.2 | NP_851596.1 | 6 | 892 | ||
| GenBank mRNA | AY258290.1 | - | 6 | 892 | ||