siRNA Details
| Gene Symbol: Ms4a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106338.1 | NP_001099808.1 | 4 | 767 | ||
| XM_008760256.1 | XP_008758478.1 | 5 | 673 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ms4a7| Gene Symbol: Ms4a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106338.1 | NP_001099808.1 | 4 | 767 | ||
| XM_008760256.1 | XP_008758478.1 | 5 | 673 | |||