siRNA Details
| Gene Symbol: Ms4a6bl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006975.1 | NP_001006976.1 | 5 | 614 | ||
| GenBank mRNA | BC083691.1 | - | 5 | 614 | 7189412 | |
| FQ230476.1 | - | 618 | ||||
| Gene Symbol: LOC100912573 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231109.2 | XP_006231171.1 | 666 | |||

Ms4a6bl
Genomic Map