siRNA Details
Gene Symbol: Ms4a6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231110.2 | XP_006231172.1 | 5 | 931 | ||
XM_215145.8 | XP_215145.5 | 5 | 661 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ms4a6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231110.2 | XP_006231172.1 | 5 | 931 | ||
XM_215145.8 | XP_215145.5 | 5 | 661 |