siRNA Details
| Gene Symbol: Mrpl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009647.1 | NP_001009647.1 | 1, 2 | 132 | ||
| GenBank mRNA | BC058154.1 | - | 1, 2 | 136 | ||
| BC088312.1 | - | 1, 2 | 132 | 7324682 | ||
| FQ212609.1 | - | 1, 2 | 142 | |||
| FQ215871.1 | - | 1, 2 | 144 | |||
| FQ219747.1 | - | 1, 2 | 130 | |||

Mrpl16
Genomic Map