siRNA Details
| Gene Symbol: Mrpl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009647.1 | NP_001009647.1 | 3 | 232 | ||
| GenBank mRNA | BC058154.1 | - | 3 | 236 | ||
| BC088312.1 | - | 3 | 232 | 7324682 | ||
| EU148065.1 | - | 2 | 63 | |||
| FQ212609.1 | - | 3 | 242 | |||
| FQ214728.1 | - | 1 | 20 | |||
| FQ215871.1 | - | 3 | 244 | |||
| FQ219747.1 | - | 3 | 230 | |||

Mrpl16
Genomic Map