siRNA Details
| Gene Symbol: Olr323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000245.1 | NP_001000245.1 | 1 | 592 | ||
| Gene Symbol: Olr322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000762.1 | NP_001000762.1 | 1 | 592 | ||
| XM_008760258.1 | XP_008758480.1 | 3 | 1146 | |||
| XM_008760259.1 | XP_008758481.1 | 4 | 1041 | |||

Olr323
Genomic Map