siRNA Details
| Gene Symbol: Dtx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047855.1 | NP_001041320.1 | 5 | 403 | ||
| XM_006231114.2 | XP_006231176.1 | 6 | 2224 | |||
| GenBank mRNA | AY383692.1 | - | 5 | 403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dtx4| Gene Symbol: Dtx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047855.1 | NP_001041320.1 | 5 | 403 | ||
| XM_006231114.2 | XP_006231176.1 | 6 | 2224 | |||
| GenBank mRNA | AY383692.1 | - | 5 | 403 | ||