siRNA Details
| Gene Symbol: Olr340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000253.1 | NP_001000253.1 | 1 | 206 | ||
| Gene Symbol: Olr343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000254.1 | NP_001000254.1 | 1 | 206 | ||

Olr340
Genomic Map