siRNA Details
| Gene Symbol: Olr380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001277.1 | NP_001001277.1 | 1 | 136 | ||
| Gene Symbol: Olr383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000262.1 | NP_001000262.1 | 1 | 136 | ||

Olr380
Genomic Map