siRNA Details
| Gene Symbol: Pnma3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106342.2 | NP_001099812.1 | 1 | 917 | ||
| XM_006229513.2 | XP_006229575.1 | 2 | 1365 | |||
| XM_006229514.2 | XP_006229576.1 | 2 | 1365 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnma3| Gene Symbol: Pnma3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106342.2 | NP_001099812.1 | 1 | 917 | ||
| XM_006229513.2 | XP_006229575.1 | 2 | 1365 | |||
| XM_006229514.2 | XP_006229576.1 | 2 | 1365 | |||