siRNA Details
| Gene Symbol: Xlr4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229523.2 | XP_006229585.1 | 5 | 823 | ||
| XM_006229524.2 | XP_006229586.1 | 5 | 838 | |||
| XM_006229527.2 | XP_006229589.1 | 5 | 443 | |||
| XM_006229528.2 | XP_006229590.1 | 5 | 805 | |||
| XM_006229529.2 | XP_006229591.1 | 5 | 820 | |||
| XM_219738.8 | XP_219738.6 | 4 | 982 | |||

Xlr4a
Genomic Map