siRNA Details
| Gene Symbol: Flna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134599.1 | NP_001128071.1 | 39 | 6738 | ||
| XM_006229569.2 | XP_006229631.1 | 41 | 6816 | |||
| XM_006229570.2 | XP_006229632.1 | 40 | 6792 | |||
| GenBank mRNA | FJ416060.1 | - | 39 | 6685 | ||
| FQ230835.1 | - | 16 | 2346 | |||

Flna
Genomic Map