siRNA Details
| Gene Symbol: Flna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134599.1 | NP_001128071.1 | 44 | 7535 | ||
| XM_006229569.2 | XP_006229631.1 | 46 | 7613 | |||
| XM_006229570.2 | XP_006229632.1 | 45 | 7589 | |||
| GenBank mRNA | FJ416060.1 | - | 44 | 7482 | ||
| FQ227568.1 | - | 2 | 222 | |||
| FQ230835.1 | - | 21 | 3143 | |||
| FQ232712.1 | - | 3 | 325 | |||
| FQ234693.1 | - | 3 | 326 | |||

Flna
Genomic Map