siRNA Details
| Gene Symbol: Fam50a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170573.1 | NP_001164044.1 | 4, 5 | 572 | ||
| GenBank mRNA | BC168887.1 | - | 2, 3 | 165 | ||
| Gene Symbol: LOC100910130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749779.3 | XP_003749827.1 | 4, 5 | 556 | ||

Fam50a
Genomic Map