siRNA Details
| Gene Symbol: Lage3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106345.1 | NP_001099815.1 | 3 | 465 | ||
| Gene Symbol: RGD1309036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228990.2 | XP_006229052.1 | 2 | 782 | ||

Lage3
Genomic Map