siRNA Details
| Gene Symbol: RGD1311863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024974.1 | NP_001020145.1 | 4 | 713 | ||
| GenBank mRNA | BC091268.1 | - | 4 | 713 | 7380414 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_026120, BC092281, BC043335, AK010784.

RGD1311863
Genomic Map