siRNA Details
| Gene Symbol: RGD1311863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024974.1 | NP_001020145.1 | 5 | 859 | ||
| GenBank mRNA | BC091268.1 | - | 5 | 859 | 7380414 | |
| FQ223854.1 | - | 1 | 129 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1311863| Gene Symbol: RGD1311863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024974.1 | NP_001020145.1 | 5 | 859 | ||
| GenBank mRNA | BC091268.1 | - | 5 | 859 | 7380414 | |
| FQ223854.1 | - | 1 | 129 | |||