siRNA Details
Gene Symbol: RGD1311863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024974.1 | NP_001020145.1 | 3 | 458 | ||
GenBank mRNA | BC091268.1 | - | 3 | 458 | 7380414 | |
FQ224897.1 | - | 3 | 493 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026120, BC092281, AI509964, BC043335, AK010784.
