siRNA Details
| Gene Symbol: Bloc1s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037349.2 | NP_001032426.1 | 4 | 405 | ||
| XM_008760400.1 | XP_008758622.1 | 4 | 432 | |||
| XM_008760401.1 | XP_008758623.1 | 5 | 507 | |||
| GenBank mRNA | AY786315.1 | - | 4 | 386 | ||
| EF061226.2 | - | 5 | 461 | |||

Bloc1s2
Genomic Map