siRNA Details
| Gene Symbol: Hoga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106355.1 | NP_001099825.1 | 4 | 695 | ||
| XM_006231386.2 | XP_006231448.1 | 3 | 693 | |||
| GenBank mRNA | FQ210513.1 | - | 4 | 509 | ||
| FQ218423.1 | - | 4 | 507 | |||

Hoga1
Genomic Map