siRNA Details
| Gene Symbol: Hoga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106355.1 | NP_001099825.1 | 2 | 435 | ||
| GenBank mRNA | FQ210513.1 | - | 2 | 249 | ||
| FQ218423.1 | - | 2 | 247 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoga1| Gene Symbol: Hoga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106355.1 | NP_001099825.1 | 2 | 435 | ||
| GenBank mRNA | FQ210513.1 | - | 2 | 249 | ||
| FQ218423.1 | - | 2 | 247 | |||