siRNA Details
| Gene Symbol: Ndufb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106360.2 | NP_001099830.1 | 4 | 335 | ||
| GenBank mRNA | BC166889.1 | - | 4 | 335 | 6891455 | |
| BC166905.1 | - | 4 | 335 | 6887475 | ||
| BC166906.1 | - | 4 | 335 | 6888064 | ||
| FQ217125.1 | - | 4 | 329 | |||
| FQ217646.1 | - | 4 | 338 | |||
| FQ223847.1 | - | 4 | 350 | |||

Ndufb8
Genomic Map