siRNA Details
| Gene Symbol: Kazald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033064.1 | NP_001028236.1 | 4 | 864 | ||
| GenBank mRNA | BC103652.1 | - | 4 | 864 | 7317353 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kazald1| Gene Symbol: Kazald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033064.1 | NP_001028236.1 | 4 | 864 | ||
| GenBank mRNA | BC103652.1 | - | 4 | 864 | 7317353 | |