siRNA Details
| Gene Symbol: Dpcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013905.1 | NP_001013927.1 | 4 | 403 | ||
| XM_006231450.2 | XP_006231512.1 | 4 | 444 | |||
| GenBank mRNA | BC078988.1 | - | 4 | 403 | 7112530 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpcd| Gene Symbol: Dpcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013905.1 | NP_001013927.1 | 4 | 403 | ||
| XM_006231450.2 | XP_006231512.1 | 4 | 444 | |||
| GenBank mRNA | BC078988.1 | - | 4 | 403 | 7112530 | |