siRNA Details
| Gene Symbol: Pprc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106363.1 | NP_001099833.1 | 9 | 3951 | ||
| XM_006231451.2 | XP_006231513.1 | 9 | 3972 | |||
| XM_006231452.2 | XP_006231514.1 | 9 | 3968 | |||
| XM_006231453.2 | XP_006231515.1 | 9 | 3965 | |||
| XM_006231454.2 | XP_006231516.1 | 9 | 3972 | |||
| XM_006231455.2 | XP_006231517.1 | 9 | 3963 | |||
| XM_006231456.2 | XP_006231518.1 | 9 | 3887 | |||

Pprc1
Genomic Map