siRNA Details
| Gene Symbol: Cuedc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079886.1 | NP_001073355.1 | 9 | 1238 | ||
| XM_006231458.2 | XP_006231520.1 | 9 | 828 | |||
| XM_006231459.2 | XP_006231521.1 | 9 | 1291 | |||
| XM_006231460.2 | XP_006231522.1 | 9 | 834 | |||
| XM_006231462.2 | XP_006231524.2 | 9 | 1647 | |||
| XM_006231465.2 | XP_006231527.1 | 9 | 774 | |||
| XM_006231466.2 | XP_006231528.1 | 9 | 780 | |||
| XM_008760408.1 | XP_008758630.1 | 9 | 1644 | |||
| XM_008760409.1 | XP_008758631.1 | 9 | 1459 | |||
| XM_008760410.1 | XP_008758632.1 | 8 | 1562 | |||
| XM_008760411.1 | XP_008758633.1 | 9 | 1684 | |||
| XM_008760412.1 | XP_008758634.1 | 10 | 877 | |||
| GenBank mRNA | BC127518.1 | - | 9 | 1238 | 7104051 | |
| FQ220195.1 | - | 9 | 742 | |||
| FQ229647.1 | - | 8 | 687 | |||

Cuedc2
Genomic Map