siRNA Details
| Gene Symbol: Actr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106364.1 | NP_001099834.1 | 4 | 305 | ||
| XM_006231470.2 | XP_006231532.1 | 4 | 477 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Actr1a| Gene Symbol: Actr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106364.1 | NP_001099834.1 | 4 | 305 | ||
| XM_006231470.2 | XP_006231532.1 | 4 | 477 | |||