siRNA Details
| Gene Symbol: Cep55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025646.1 | NP_001020817.1 | 3 | 416 | ||
| XM_006231318.2 | XP_006231380.1 | 3 | 396 | |||
| XM_006231319.2 | XP_006231381.1 | 3 | 510 | |||
| XM_006231320.2 | XP_006231382.1 | 3 | 353 | |||
| GenBank mRNA | BC098013.1 | - | 3 | 416 | 7454372 | |

Cep55
Genomic Map